TLC Analysis of Pharmaceutical Products on HPTLC Silica Gel 60 F 254
TLC Analysis of Pharmaceutical Products on HPTLC Silica Gel 60 F 254
Conditions
plate 1.05642 HPTLC pre-coated plate silica gel 60 F 254s (1.05642.0001, 1.05548.0001)
eluent [A] dichloromethan; [B] ethanol; (95:5, A:B)
migration distance 7 cm
chamber normal chamber without chamber saturation
sample 0.2 μL
detector in situ evaluation with TLC/HPTLC scanner (Camag) UV 254 nm
Description
suitability application for TLC