TLC Analysis of Phenol Carboxylic Acids on HPTLC Silica Gel 60 F 254 with Concentrating Zone
TLC Analysis of Phenol Carboxylic Acids on HPTLC Silica Gel 60 F 254 with Concentrating Zone
Conditions
plate HPTLC pre-coated plate silica get 60F 254 with concentrating zone
(1.13728.0001, 1.13727.0001)
eluent [A] acetonitrile; [B] water; (95:5, A:B)
migration distance 6 cm
chamber normal chamber without chamber saturation
sample 1 μL
detector in situ evaluation with TLC/HPTLC scanner (Camag) UV 366 nm
Description
Featured Industry Pharmaceutical (small molecule)
suitability application for TLC