TLC Analysis of Phospholipids on HPTLC LiChrospher Silica gel 60 F254s
TLC Analysis of Phospholipids on HPTLC LiChrospher Silica gel 60 F254s
Conditions
plate 1.15445 HPTLC Silica gel 60 RP-18 F 254s MS-grade 20cm x 10 cm (1.15445.0001)
eluent [A] isopropanol; [B] methanol; [C] aceton; [D] water; (20:15:5:1, A:B:C:D)
migration distance 5 cm
chamber normal chamber without chamber saturation
detector Visible light
Description
suitability application for TLC
Featured Industry Chemicals and Industrial Polymers