Separation of plant pigments on Silica gel 60 HPTLC plates
Separation of plant pigments on Silica gel 60
Chromatographic Conditions
Plate HPTLC Silica gel 60 F254, 10x10cm
Article number 1. 05628
Mobile Phase (v/v) n-heptane/1-propanol (9/1)
Migration distance 5cm
Application volume 5 µL
sample preparation milling leafs in a mortar with acetone
Chamber normal chamber without chamber saturation
Detection white light
Chromatographic Data
No.   Compounds Solvent
1 to 6 leafs carotenes acetone
    pheophytin  
    chlorophyll a  
    chlorophyll b  
    lutein  
    violaxanthin  
    neoxanthin  
Materials