TLC Analysis of Plasticizers on HPTLC Silica Gel 60 F 254 with Concentrating Zone
TLC Analysis of Plasticizers on HPTLC Silica Gel 60 F 254 with Concentrating Zone
Conditions
plate TLC pre-coated plate silica gel 60 F 254s with concentrating zone (1.13728.0001, 1.13727.0001)
eluent [A] petroleum benzene 40-60 °C; [B] acetone; (90:10, A:B)
migration distance 5 cm
chamber normal chamber without chamber saturation
sample 1 μL
detector in situ-evaluation with TLC/HPTLC scanner (Camag) UV 265 nm
Description
suitability application for TLC
Featured Industry Chemicals and Industrial Polymers