TLC Analysis of Polystyrene 800 on TLC pre-coated plate silica gel 60
TLC Analysis of Polystyrene 800 on TLC pre-coated plate silica gel 60
Conditions
plate TLC pre-coated plate silica gel 60 F 254 with concentrating zone
Mobile phase carbon tetrachloride
migration distance 7 cm
chamber normal chamber without chamber saturation
detection in-situ evaluation with TLC/HPTLC scanner (Camag)
UV 254 nm
Description
Application No. TLC2018297
Featured Industry Chemicals and Industrial Polymers 