TLC Analysis of Polyvalent Phenols on HPTLC NH2 F254s (analyte set 2)
TLC Analysis of Polyvalent Phenols on HPTLC NH2 F254s (analyte set 2)
Conditions
plate HPTLC pre-coated plate NH2 F 254s (1.13192.0001, 1.15647.0001)
eluent [A] chloroform; [B] methanol; (50:50, A:B)
migration distance 7 cm
chamber normal chamber without chamber saturation
sample 0.2 μL
detector in situ-evaluation with TLC/HPTLC scanner (Camag) UV 254 nm
Description
Featured Industry Chemicals and Industrial Polymers
suitability application for TLC