TLC Analysis of Progesterones on HPTLC CN F254s
TLC Analysis of Progesterones on HPTLC CN F254s
Conditions
plate TLC pre-coated plate CN F 254s (1.16464.0001)
eluent A] petroleum benzine (40-60°); [B] acetone; [C] water;
(a. straight-phase system: (80:20, A:B) b. reversed-phase system: (60:40, B:C) )
migration distance 7 cm
chamber normal chamber without chamber saturation
sample 0.2 μL
detector in situ-evaluation with TLC/HPTLC scanner (Camag) UV 248 nm
Description
Featured Industry Life Science and Biopharma
suitability application for TLC