TLC Analysis of Progesterones on HPTLC DIOL F254s (analyte set 3)
TLC Analysis of Progesterones on HPTLC DIOL F254s (analyte set 3)
Conditions
plate HPTLC pre-coated plate DIOL F 254s (1.12668.0001)
eluent [A] petroleum benzine (40-60°); [B] acetone; (75:25, A:B)
migration distance 7 cm
chamber normal chamber without chamber saturation
sample 0.3 μL
detector Spray reagent: MnCl2-sulfuric acid heated up to 110° C for 5 min (in situ-evaluation with TLC/HPTLC scanner (Camag) UV 366 nm)
Description
suitability application for TLC
application for TLC Life Science and Biopharma