TLC Analysis of Protected Tetrapeptides on HPTLC NH2 F254s
TLC Analysis of Protected Tetrapeptides on HPTLC NH2 F254s
Conditions
plate TLC pre-coated plate NH2 F 254s (1.13192.0001, 1.15647.0001 )
eluent [A] toluene; [B] aceton; [C] methanol; (50:40:10, A:B:C) including 0.1 M lithium chloride
migration distance 8 cm
chamber normal chamber without chamber saturation
detector dipped in dichlorofluorescein (0.2% in ethanol) (Spray reagent: 5% perchloric acid in methanol heating up to 120°C for 15 min)
Description
Featured Industry Life Science and Biopharma
suitability application for TLC