TLC Analysis of Pyrimidine Derivatives on HPTLC Silica Gel 60 F 254 with Concentrating Zone (analyte set 2)
TLC Analysis of Pyrimidine Derivatives on HPTLC Silica Gel 60 F 254 with Concentrating Zone (analyte set 2)
Conditions
plate TLC pre-coated plate silica gel 60 F 254 with concentrating zone
(1.13728.0001, 1.13727.0001)
eluent [A] chloroform; [B] methanol; (90:10, A:B)
migration distance 5 cm
chamber normal chamber without chamber saturation
sample 2 μL
detector in situ evaluation with TLC/HPTLC scanner (Camag) UV 265 nm