TLC Analysis of Salicylic Acid on HPTLC Silica 60F 254s
TLC Analysis of Salicylic Acid on HPTLC Silica 60F 254s >
Conditions
plate HPTLC pre-coated plate silica gel 60 F 254s extra pure (suitable for aqueous eluents)
eluent
chloroform/acetonitrile 90/10 (v/v)
migration distance
7 cm
chamber normal chamber without chamber saturation
detection in-situ evaluation with TLC/HPTLC scanner (Camag) UV 254 nm
Description
Featured Industry Life Science and Biopharma 
Analysis Note 0
Application No. BOR190918C