TLC Analysis of Solenasol on Silica Gel 60
TLC Analysis of Solenasol on Silica Gel 60
Conditions
plate HPTLC pre-coated plate silica gel 60 F 254s 20x10 (Product No. 1.05642)
Mobile phase n-heptane/ethylacetate (80/20)
migration distance each 7 cm
chamber normal chamber without chamber saturation
detection 20% perchloric acid in ethanol spray 20s and heat 5min at 100°C, Detection white light
Description
Application No. BOR200918C
Featured Industry Vitamins, Nutraceuticals, Herbal Medicines
Materials
Related Links
Categories