TLC Analysis of Some Benzenecarboxylic Acids on HPTLC DIOL F254s
TLC Analysis of Some Benzenecarboxylic Acids on HPTLC DIOL F254s
Conditions
plate  1.13192 HPTLC pre-coated plate NH2 F 254s (1.13192.0001, 1.15647.0001)
eluent [A] ethanol; [B] ammonia pH 12; (60:40, A:B) containing 0.18 M NaCl
migration distance  5 cm
chamber  normal chamber without chamber saturation
sample  0.3 μL
detector  in situ evaluation with TLC/HPTLC scanner (Camag) UV 254 nm.
Descriptions
suitability application for TLC
Featured Industry Chemicals and Industrial Polymers