TLC Analysis of some dansyl amino acids on a polyamide pre-coated plate
TLC Analysis of some dansyl amino acids on a polyamide pre-coated plate
Conditions
plate TLC pre-coated plate polyamide 11 F 254
Mobile phase methanol/acetic acid/water 80/10/10 (v/v)
migration distance 10 cm
chamber normal chamber without chamber saturation
sample application volume 500 nl
detection in-situ evaluation with TLC/HPTLC scanner (Camag)
UV 366 nm
Description
Application No. TLC2018287