TLC Analysis of Some Pesticides on HPTLC DIOL F254s
TLC Analysis of Some Pesticides on HPTLC DIOL F254s
Conditions
plate 1.05636 HPTLC pre-coated plate DIOL F 254 (1.05636.0001, 1.12668.0001)
eluent [A] petroleum benzine (40 - 60°); [B] acetone; (90:10, A:B)
migration distance 7 cm
chamber normal chamber without chamber saturation
sample 0.3 μL
detector in situ evaluation with TLC/HPTLC scanner (Camag) UV 238 nm
Description
suitability application for TLC
Featured Industry Environmental