TLC Analysis of Some Pesticides on HPTLC pre-coated plate DIOl F 254s
TLC Analysis of Some Pesticides on HPTLC pre-coated plate DIOl F 254s
Conditions
plate HPTLC pre-coated plate DIOL F 254s
eluent petroleum benzine (40 - 600)/acetone 85/15 (v/v)
migration distance 7 cm
chamber normal chamber without chamber saturation
sample application volume 300 nL
detection  in situ-evaluation with TLC/HPTLG scanner (Camag) UV 230 nm
Description
Featured Industry Environmental 
Application No. BOR160918B