TLC Analysis of Steroids on HPTLC Silica gel 60 DIOL F254s
TLC Analysis of Steroids on HPTLC Silica gel 60 DIOL F254s
Conditions
plate HPTLC Silica gel 60 DIOL F254s 20x10cm (Product No. 1.05636)
Mobile phase petroleum benzine 40-60°C/acetone (80/20)
migration distance 5 cm
chamber normal chamber without chamber saturation
sample application volume 1 µl
detection 20% perchloric acid in water spray 20s and heat 5min at 100°C, Detection white light
Description
Application No. BOR050918C