TLC Analysis of Sugars on HPTLC Silica gel 60 NH2
TLC Analysis of Sugars on HPTLC Silica gel 60 NH2
Conditions
plate HPTLC Silica gel 60 NH2 20x10cm (Product No. 1.13192)
Mobile phase 1. water/ACN (30/70)  2. ethylacetate/pyridine/water/glacial acetic acid/propanoic acid (50/50/10/5/5)
migration distance 1. 55 mm 2. 75 mm
chamber normal chamber without chamber saturation
sample application volume 2 µl
detection heat 5 min at 150°C, Detection
Description
Application No. BOR210918
Featured Industry Food & Beverage