TLC Analysis of Testosterones on HPTLC DIOL F254s (analyte set 3)
TLC Analysis of Testosterones on HPTLC DIOL F254s (analyte set 3)
Conditions
plate HPTLC pre-coated plate DIOL F 254s (1.12688.0001)
eluent di-iso-propyl ether
migration distance 7 cm
chamber normal chamber without chamber saturation
sample 0.2 μL
detector Spray reagent: MnCl2-sulfuric acid heated up to 110° C for 10 min (in situ-evaluation with TLC/HPTLC scanner (Camag) UV 540 nm)
Description
Featured Industry Life Science and Biopharma
suitability application for TLC