TLC analysis of tri-peptides TLC pre-coated plate RP-18 F 254s
TLC analysis of tri-peptides TLC pre-coated plate RP-18 F 254s
plate  HPTLC pre-coated plate RP-18 F 254s
eluent Acetonitrile/buffer solution pH 3.00 90/10 (v/v), with addition of 0,1 mol/L lithium chloride
migration distance 7 cm
chamber normal chamber without chamber saturation
sample application volume  
detection spray reagent: ninhydrin heating up to 110 C for 20 min.
Description
Featured Industry Life Science and Biopharma 
Application No. BOR170918A