TLC Analysis of Triazines on HPTLC NH2 F254s
TLC Analysis of Triazines on HPTLC NH2 F254s
Conditions
plate  HPTLC pre-coated plate NH2 F 254s (1.13192.0001, 1.15647.0001)
eluent [A] chloroform; [B] toluene; (90:10, A:B)
migration distance  7 cm
chamber  normal chamber without chamber saturation
sample  0.3 μL
detector  in situ-evaluation with TLC/HPTLC scanner (Camag) UV 238 nm
Description
suitability application for TLC