TLC Analysis of Tripeptides on HPTLC Silica Gel 60 RP-18 F254s
TLC Analysis of Tripeptides on HPTLC Silica Gel 60 RP-18 F254s
Conditions
chamber normal chamber without chamber saturation A043
eluent [A] acetonitrile; [B] buffer solution, pH 3.00; (90:10, A:B)
containing 0.1 M lithium chlorideA041
plate TLC pre-coated plate RP-18 F 254s
migration distance 7cm
detector spray reagent: ninhydrin, heated to 110 °C for 20 min
Description
Categories Analytical Chromatography, TLC Applications, steroids
Featured Industry Life Science and Biopharma
General description TLC Plate Information: 1.13724.0001
suitability application for TLC