TLC Analysis of Uridine and Phosphate Analogs on HPTLC NH2 F254s
TLC Analysis of Uridine and Phosphate Analogs on HPTLC NH2 F254s
Conditions
plate HPTLC pre-coated plate NH2 F 254s (1.13192.0001, 1.15647.0001)
eluent [A] acetone; [B] water; (30:70, A:B) containing 0.2 M NaCl
migration distance 5 cm
chamber normal chamber without chamber saturation
detector in situ evaluation with TLC/HPTLC scanner (Camag) UV 254 nm
Description
Featured Industry Life Science and Biopharma
suitability application for TLC