TLC Analysis of Uridine and Phosphate Analogs on HPTLC pre-coated plate PEl-Cellulose F
TLC Analysis of Uridine and Phosphate Analogs on HPTLC pre-coated plate PEl-Cellulose F
Conditions
plate 1.05725 HPTLC pre-coated plate PEI-Cellulose F (1.05725.0001, 1.05579.0001)
eluent Acetic acid 1N/ethanol 80/20 (v/v) with addition of 0.5 mol/l lithium chloride
migration distance 7 cm
chamber normal chamber without chamber saturation
sample 0.4 μL
detector in situ evaluation with TLC/HPTLC scanner (Camag) UV 270 nm
Description
suitability application for TLC