Separation of UV-Filter on Silica gel 60 HPTLC plates
Separation of UV-Filter on Silica gel 60
Eusolex® HMS
Eusolex® HMS
Eusolex® 9020
Eusolex® 9020
Chromatographic Conditions
Plate HPTLC Silica gel 60 F254s 20x10cm
Article number 1. 15696
Mobile Phase (v/v) n-heptane/tetrahydrofuran (4/1)
Migration distance 50mm
Application volume 1 µL
Chamber normal chamber without chamber saturation
Detection Detection UV @ 254nm
Chromatographic Data
No. Compounds       Concentration (mg/ml) Solvent hRf
1 and 10 Eusolex® HMS 0.4 ethanol 84
2 and 11 Eusolex® HMS 1.0 ethanol 84
3 and 4 Eusolex® HMS, Eusolex 9020,
Oxynex ST and
Ronacare AP sample 1
- ethanol -
5 and 14 Eusolex® HMS 1.5 ethanol 84
6 and 15 Eusolex® HMS 2.0 ethanol 84
7 and 16 Eusolex® HMS 3.0 ethanol 84
8 and 9 Eusolex® HMS, Eusolex 9020,
Oxynex ST and
Ronacare AP sample 1
- ethanol -
12 and 13 Eusolex® HMS, Eusolex 9020,
Oxynex ST and
Ronacare AP sample 1
- ethanol -
Materials