Separation of UV-Filter on Silica gel 60 RP-18 HPTLC plates
TLC Application number 338
Separation of UV-Filter on Silica gel 60 RP-18
Eusolex® OCR
Eusolex® OCR
Eusolex® 9020
Eusolex® 9020
Chromatographic Conditions
Plate HPTLC Silica gel 60 RP-18 F254s 20x10cm
Article number 1. 16225
Mobile Phase (v/v) methanol/acetonitrile (90/10)
Migration distance 50mm
Application volume 1 µL
Chamber normal chamber without chamber saturation
Detection Detection UV @ 254nm
Chromatographic Data
No. Compounds Concentration (mg/ml) Solvent hRf
1 and 10 Eusolex® 9020 0.1 ethanol 49
2 and 11 Eusolex® 9020 0.2 ethanol 49
3 and 4 Eusolex® OCR and Eusolex® 9020, sample 1 -
ethanol -
5 and 14 Eusolex® 9020 0.4 ethanol 49
6 and 15 Eusolex® 9020 0.8 ethanol 49
7 and 16 Eusolex® 9020 2.0 ethanol 49
8 and 9 Eusolex® OCR and Eusolex® 9020, sample 2 - ethanol -
12 and 13 Eusolex® OCR and Eusolex® 9020, sample 3 - ethanol -
Materials