TLC Analysis of Vitamins on HPTLC NH2 F254s
TLC Analysis of Vitamins on HPTLC NH2 F254s
Conditions
plate: HPTLC pre-coated plate NH2 F 254s (1.13192.0001, 1.15647.0001)
eluent: [A] acetonitrile; [B] water; (70:30, A:B)
migration distance: 7 cm
chamber: normal chamber without chamber saturation
sample 0.3 μL
detector (in situ evaluation with TLC/HPTLC scanner (Camag) UV 280 nm )
Description
Featured Industry Vitamins, Nutraceuticals, and Natural Products
suitability application for TLC