TLC Analysis of Xanthine and its N-methyl Derivatives on HPTLC NH2 F254s
TLC Analysis of Xanthine and its N-methyl Derivatives on HPTLC NH2 F254s
Conditions
plate HPTLC pre-coated plate NH2 F 254s (1.13192.0001, 1.15647.0001)
eluent methanol
migration distance 5 cm
chamber normal chamber without chamber saturation
sample 0.1 μL
detector in situ-evaluation with TLC/HPTLC scanner (Camag) UV 254 nm
Description
Featured Industry Life Science and Biopharma
suitability application for TLC