TLC Analysis Ofacetylsalicylic Acid in Painkillers on HPTLC Silica gel 60 F254
TLC Analysis Ofacetylsalicylic Acid in Painkillers on HPTLC Silica gel 60 F254
Conditions
sample preparation 1 tablet was dissolved in 10 mL water and filtered over a 0.45 μm syringe filter.
Application by CAMAG ATS4 6 mm bandwise.
plate 1.05628 HPTLC Silica gel 60 F254 10 cm x10 cm (1.05628.0001)
eluent [A] ethylacetate; [B] methanol; (4 :1, A:B)
migration distance 5 cm
chamber normal chamber without chamber saturation
detector 366 nm
Description
suitability application for TLC