Sample prep-free determination of Arginine and Taurine in shampoo by using TLC-MS
The application of modern thin-layer chromatography (TLC) offers a lot of advantages especially in the field of cosmetics. The ingredients can usually be separated directly from complex matrices and many samples can be analysed quickly and simultaneously under exactly the same conditions.
Below the analysis of taurine and arginine in shampoo is described. Because of the high matrix tolerance of TLC, only a minimum of sample preparation is necessary to detect the analytes in shampoo. Quantification is shown by densitometry and straight forward mass analysis is performed by coupling the TLC-MS Interface from Camag with an ESI mass detector.
The developed TLC plates were treated with ninhydrin spray reagent to derivatise arginine and taurine and make it visible under white light. While taurine was observed at an hRf value of 24, arginine showed no mobility under the used conditions and was located at an hRf value of 0. Therefore only taurine was used for quantitative evaluation. By using the peak area, a triple determination of the shampoo sample and a 3-point calibration, the amount of taurine was determined to be 75 mg, based on the amount of cosmetic formulation used. This corresponds to the usual amount of 0.1% active ingredient in a formulation. Chromatographically separated TLC plate is shown after derivatisation in Figure 1a. By comparison of the shampoo sample to standards, band positions of arginine and taurine could be determined. Figure 1b shows the observed arginine and taurine signals on the TLC scan at 600 nm examining track 6.
TLC plate under white light, 1b: TLC scan at 600 nm, chromatographic data shown on next page
Figure 1a: TLC plate under white light, 1b: TLC scan at 600 nm, chromatographic data shown on next page.
For assignment of the bands of interest mass spectrometry with an mass detector was performed after elution of the sample zone with the TLC-MS Interface. The mass spectra show the molecules at m/z 175.2 for arginine (Fig. 2) and m/z 124.1 for taurine (Fig. 3).
Mass spectrum and structure of arginine
Figure 2: Mass spectrum and structure of arginine, recorded at an hRf value of 0 at track 4, measured with positive ionization.
Mass spectrum and structure of taurine
Figure 3: Mass spectrum and structure of taurine, recorded at an hRf value of 24 at track 4, measured with negative ionization.
These results demonstrate that arginine and taurine can easily be measured by TLC-MS without the need of complex sample preparation.
Application data
Plate HPTLC Silica gel 60 WR F254s, 20x10 cm
Sample preparation 15 min. stirring in 2-Propanol, filtration
Sample application ATS 4 sample applicator (Camag), 6 mm bandwise
Application volume 0.5 – 5 μL
Mobile phase 2-Propanol / Water (8/2 v/v)
Migration distance 5 cm
Migration time 50 min
Extraction equipment “TLC-MS Interface” from Camag
Extraction solvent Acetonitrile / Water (95/5 v/v) + 0.1% Formic acid
Extraction flow 0.2 mL/min
Documentation equipment documentation unit Reprostar / Digistore (Camag)
Wavelength white light, 600 nm
Staining none
MS equipment single-quadrupole mass spectrometer
MS detection ESI (+/-) mode MS (m/z 100 – 500)
Chromatographic data
Track Compound Conc. [mg/mL] Application volume [μL] hRf Detected mass m/z
01-Mar Arginine standard 0.1 0.5, 1.0, 2.0 0 175.2
04-Jun Shampoo sample - 5 0/24 175.2 / 124.1
07-Sep Taurine standard 0.1 0.5, 1.0, 2.0 24 124.1
Ordering information
Description Product No.
HPTLC Silica gel 60 F254 MS-grade, 20x10 cm 1.00934
Acetonitrile hypergrade for LC-MS LiChrosolv® 1.00029
2-Propanol gradient grade for liquid chromatography LiChrosolv® 1.0104
Formic acid for analysis EMSURE® 1.00264
Ninhydrin spray solution for thin-layer chromatography 1.06705
Water for chromatography (LC-MS Grade) LiChrosolv® 1.15333
Millex®-FH filter, 0.45 μm hydrophobic PTFE, 25 mm, non-sterile SLFH025NS