TLC Analysis of Tri-peptides on HPTLC pre-coated plate RP-8 F 254s
TLC Analysis of Tri-peptides on HPTLC pre-coated plate RP-8 F 254s
Conditions
plate HPTLC pre-coated plate RP-8 F 254s
eluent acetonitrile/buffer solution pH 3.00 90/10 with addition of 0,1 mo1/1 lithium chloride
migration distance 5 cm
chamber normal chamber without chamber saturation
detection spray reagent: ninhydrin heating up to 110 C for 20 min.
Description
Application No. BOR180918B
Featured Industry Life Science and Biopharma
Compound Class Peptides