Unmodified Silica TLC Plates
TLC plates manufactured by Merck KGaA, Darmstadt Germany allow reliable, fast and economical analysis of a great range of substances in pharmaceutical, environmental, food and beverage, cosmetics and many more applications. Thanks to a highly standardized production process and an outstanding quality control, our TLC plates offer the highest batch-to-batch reproducibility. This ensures highly reliable results in qualitative as well as in quantitative analysis.
Excellent Separation Efficiency for a Broad Range of Substances
Classical silica TLC plates enable the analysis of a broad range of compounds and cover almost 80% of applications for adsorption and partition thin-layer chromatography. The TLC plates supplied by Merck KGaA Darmstadt, are based on our proven silica gel 60 combined with a unique polymeric binder, resulting in a uniform and hard surface that will not crack or blister. The smooth and extremely dense surface coating ensures narrow chromatographic bands for maximum separation performance. This is of particular importance for applications such as scanning densitometry.
Classical silica TLC plates have a main particle size distribution of 10 to 12 μm and are available with several backings in a wide range of plate dimensions. The plates have a layer thickness of 250 μm for glass, or 200 μm for aluminum or plastic sheets.
For UV detection of colorless substances, our classical silica TLC plates are available with two types of inorganic fluorescent indicators: green fluorescing F254, or blue fluorescing, acid-stable F254s.
Features and benefits
  • Suitable for a broad range of TLC applications
  • Extremely robust surface will not crack or blister
  • Available as glass, aluminum or plastic in various sizes
  • Offered with or without fluorescence indicators F254 or F254s
Application areas
Classical silica TLC plates are particularly suitable for:
  • In-process controls in drug synthesis
  • Identity and stability testing of drugs
  • Quality control of pharmaceuticals, food and environmental compounds
  • Residue analysis in food and environmental samples
Materials
Product No. Description Layer Sorbent/Coating Material Layer Thickness/ µm Format/ cm Support/ Backing Pack size/ content
1.05721.0001 TLC Silica gel 60 Silica gel 60 250 20 x 20 glass 25 plates
1.05626.0001 TLC Silica gel 60 Silica gel 60 250 10 x 20 glass 50 plates
1.05724.0001 TLC Silica gel 60 Silica gel 60 250 5 x 20 glass 100 plates
1.15326.0001 TLC Silica gel 60 Silica gel 60 250 2.5 x 7.5 glass 100 plates
1.05715.0001 TLC Silica gel 60 F254 Silica gel 60 F254*1 250 20 x 20 glass 25 plates
1.05729.0001 TLC Silica gel 60 F254 Silica gel 60 F254*1 250 10 x 20 glass 50 plates
1.05714.0001 TLC Silica gel 60 F254 Silica gel 60 F254*1 250 5 x 20 glass 100 plates
1.05808.0001 TLC Silica gel 60 F254 Silica gel 60 F254*1 250 5 x 20 glass 25 plates
1.05719.0001 TLC Silica gel 60 F254 Silica gel 60 F254*1 250 5 x 10 glass 200 plates
1.05789.0001 TLC Silica gel 60 F254 Silica gel 60 F254*1 250 5 x 10 glass 25 plates
1.05794.0001 TLC Silica gel 60 F254 Silica gel 60 F254*1 250 2.5 x 7.5 glass*3 100 plates
1.15327.0001 TLC Silica gel 60 F254 Silica gel 60 F254*1 250 2.5 x 7.5 glass*4 100 plates
1.15341.0001 TLC Silica gel 60 F254 Silica gel 60 F254*1 250 2.5 x 7.5 glass 500 plates
1.16485.0001 TLC Silica Gel 60 W F254s Silica Gel 60 W F254s*2 250 20 x 20 glass 25 plates
1.05805.0001 LuxPlate® silica gel 60 F254 LuxPlate® silica gel 60 F254*1 250 20 x 20 glass 25 plates
1.05804.0001 LuxPlate® silica gel 60 F254 LuxPlate® silica gel 60 F254*1 250 10 x 20 glass 50 plates
1.05802.0001 LuxPlate® silica gel 60 F254 LuxPlate® silica gel 60 F254*1 250 5 x 10 glass 25 plates
1.05801.0001 LuxPlate® silica gel 60 F254 LuxPlate® silica gel 60 F254*1 250 2.5 x 7.5 glass 100 plates
1.05553.0001 TLC Silica gel 60 Silica gel 60 200 20 x 20 aluminum 25 sheets
1.16835.0001 TLC Silica gel 60 Silica gel 60 200 5 x 10 aluminum 50 sheets
1.16487.0001 TLC Silica gel 60 W Silica gel 60 W*6 200 20 x 20 aluminum 25 sheets
1.05554.0001 TLC Silica gel 60 F254 Silica gel 60 F254*1 200 20 x 20 aluminum 25 sheets
1.05570.0001 TLC Silica gel 60 F254 Silica gel 60 F254*1 200 10 x 20 aluminum 25 sheets
1.05534.0001 TLC Silica gel 60 F254 Silica gel 60 F254*1 200 5 x 20 aluminum 100 sheets
1.16834.0001 TLC Silica gel 60 F254 Silica gel 60 F254*1 200 5 x 10 aluminum 50 sheets
1.05549.0001 TLC Silica gel 60 F254 Silica gel 60 F254*1 200 5 x 7.5 aluminum 20 sheets
1.05562.0001 TLC Silica gel 60 F254 Silica gel 60 F254*1 200 500 x 20 aluminum 1 roll
1.16484.0001 TLC Silica Gel 60 W F254s Silica Gel 60 W*6 F254s*2 200 20 x 20 aluminum 25 sheets
1.05748.0001 TLC Silica gel 60 Silica gel 60 200 20 x 20 plastic 25 sheets
1.05735.0001 TLC Silica gel 60 F254 Silica gel 60 F254*1 200 20 x 20 plastic 25 sheets
1.05750.0001 TLC Silica gel 60 F254 Silica gel 60 F254*1 200 4 x 8 plastic 50 sheets
1.05749.0001 TLC Silica gel 60 F254 Silica gel 60 F254*1 200 500 x 20 plastic 1 roll
60778-25EA Silica gel on TLC Al foil Silica gel 60 F254*1 200 20x20 aluminum 25 sheets
56524-25EA Silica gel on TLC Al foil, with fluorescent indicator Silica gel 60 F254*1, outstanding wettablility 200 20x20 aluminum 25 sheets
60800-20EA Silica gel on TLC Al foil, with fluorescent indicator Silica gel 60 F254*1 200 10x 20 aluminum 20 sheets
60805-25EA Silica gel on TLC Al foil Silica gel 60 200 20x20 aluminum 25 sheets
60805-50EA Silica gel on TLC Al foil Silica gel 60 F254*1 200 4 x 8 aluminum 50 sheets
91835-50EA Silica gel on TLC foil Silica gel 60 F254*1 200 5 x 10 aluminum 50 sheets
60763-50EA Silica gel on TLC plates Silica gel 60 F254*1 250 5 x 10 glass 50 plates
99569-100EA Silica gel on TLC plates Silica gel 60 F254*1 250 5 x 10 glass 100 plates
99570-25EA Silica gel on TLC plates Silica gel 60 250 20 x 20 glass 25 plates
99571-25EA Silica gel on TLC plates Silica gel 60 F254*1 250 20 x20 glass 25 plates
Z122726-25EA Sigma-Aldrich TLC plates Silica gel 60 F254*1 250 20 x 20 glass 25 plates
Z122785-25EA Sigma-Aldrich TLC plates Silica gel 60 F254*1 200 20 x 20 plastic 25 sheets
Z265500-1PAK Analtech TLC Uniplates TM: silical gel matrix Silica gel 60 F254*1, with organic binder 250 20 x 20 glass 25 plates
Z274283-1PAK Analtech TLC Uniplates TM: silical gel matrix Silica gel 60 F254*1,scored binder 250 10 x 20 glass 25 plates
Z292974-1PAK TLC Glass Plates   Silica gel 60 250 20 x 20 glass 25 plates
Z292990-1PAK TLC Glass Plates   Silica gel 60 F254*1 250 5 x 10 glass 200 plates
Z292990-1PAK TLC Glass Plates   Silica gel 60 F254*1 250 10 x 20 glass 50 plates
Z292990-1PAK TLC Glass Plates   Silica gel 60 F254*1 250 20 x 20 glass 25 plates
Z740213-100EA TLC Glass Plates   Silica gel 60 F254*1 250 2.5 x 7.5 glass 100 plates
Z740226-25EA TLC Aluminum Sheets   Silica gel 60 F254*1 200 20 x 20 aluminum 25 sheets
Z740235-25EA TLC Plastic Sheets   Silica gel 60 F254*1 200 20 x 20 plastic 25 sheets
99577-25EA Silica gel on TLC-PET foil Silica gel 60 F254*1 200 20 x 20 PET 25 sheets
Z740230-25EA TLC Aluminum Sheets   Silica gel 60 140 - 160 20 x20 aluminum 25 sheets
*1 F254 Fluorescent indicator
*2 F254s Fluorescent indicator, acid stable
*3 paper box
*4 plastic box W: water-resistant