Amphetamines-weight loss drugs
Sigma-Aldrich's Cerilliant brand provides the widest selection of Certified Spiking Solutions® for amphetamines and weight loss drugs including native and stable-labeled analogs of amphetamine, methamphetamine, phenyl and benzyl piperazines, MDA, MDMA, ephedrine, cathinones and bath salts. These certified reference solutions are gravimetrically prepared using precision balances that have been qualified to ensure minimal uncertainty, flame sealed under argon into ampoules for long term shelf life and are rigorously tested to ensure accuracy and shelf life through accelerated and real time studies.
Cerilliant reference solutions are manufactured and certified to the highest industry standards including ISO Guide 34, ISO 13485 and ISO/IEC 17025. Each certified reference standard is supported by a comprehensive Certificate of Analysis (COA) that provides all analytical data and uncertainty and traceability information to support regulatory requirements. Our COA includes analytical data on homogeneity and accuracy of concentration by comparison to an independently prepared calibration curve.
Related Links