Characterization of clinically relevant stable isotope-labeled recombinant proteins used as internal standards in quantitative MS workflows
By: Kevin Ray, Pegah Jalili, James Walters, David Rhee, St. Louis, MO
Objective
Characterization of heavy (SIL) proteins for use in quantitative MS workflows
Introduction
Methods for quantification of clinical protein biomarkers by mass spectrometry demand a high level of analytical quality. Procedures must be developed to minimize and control experimental variations in all steps of the workflow including protein extraction, fractionation, enrichment, proteolysis and analysis. To this end, we have characterized stable isotope labeled (SIL) full length proteins for use as internal standards in quantititative MS workflows. These SIL proteins can be spiked at the initial stage of the analytical workflow to significantly minimize variations. Clinically relevant SIL proteins expressed in E. coli, such as IGF1, and in mammalian HEK293 cells, such as thyroglobulin and APOA1, have been characterized in this study.
SIL Incorporation of ApoA1 and Thyroglobulin
Incorporation of 13C6 15N2 labeled lysine in DYVSQFEGSALGK liberated from SIL-APOA1
Figure 1. Incorporation of 13C6 15N2 labeled lysine in DYVSQFEGSALGK liberated from SIL-APOA1 produced by us and another commercial source.
Incorporation of 13C6 15N4 labeled arginine in two surrogate peptides
Figure 2. Incorporation of 13C6 15N4 labeled arginine in two surrogate peptides liberated from SIL-Thyroglobulin.
15N-Incorporation Analysis of SIL-IGF1
Experimentally measured and predicted isotope distributions of GFYFNKPTGYGSSSR of SIL-IGF1
Figure 3. Experimentally measured and predicted isotope distributions of GFYFNKPTGYGSSSR of SIL-IGF1. The % incorporation of 15N atoms was calculated to be 99.0% using the best correlation between experimentally measured isotope distribution and predicted isotope distributions spanning 95- 100% theoretical incorporation.
Purity Analysis of SIL-IGF1
RP-LC-UV analysis of intact SIL-IGF1 at 214 nm wavelength
Figure 4. RP-LC-UV analysis of intact SIL-IGF1 at 214 nm wavelength. A significant amount of disulfide-swapped isomer, which elutes earlier in RP-LC, was observed in a commercially sourced SIL-IGF1.
Intact Mass Analysis of SIL-ApoA1
Intact spectrum of SIL-APOA1 reveals a mixture of the proprotein (+RHFWQQ) and fully mature protein
Figure 5. Intact spectrum of SIL-APOA1 reveals a mixture of the proprotein (+RHFWQQ) and fully mature protein.
LC-HRMS of Intact IGF1 in Human Serum
SIL-IGF1 spiked into human serum fortified with 5 g/mL light IGF1
Figure 6. SIL-IGF1 was spiked into human serum fortified with 5 g/mL light IGF1. Proteins were extracted using acetone precipitation and analyzed using LC-HRMS. The most abundant isotopes of the most abundant charge state were used to create extracted ion chromatograms (not shown) for quantitation.
SIL-APOA1 Digestion in Human Sera
sil-apoa1 digestion if human sera
Figure 7. SIL-APOA1 was spiked into human serum, denatured with urea, digested using FASP protocol, and sampled over a 24-hour digestion timecourse.
Summary
We have characterized stable isotope labeled (SIL) full length versions of IGF1, Thyroglobulin, and APOA1 for utility as internal standards in quantititative MS workflows. These clinically-relevant SIL proteins can be spiked at the initial stage of the analytical workflow to significantly minimize process variations and therefore represent a more suitable choice for use as an internal standard in quantitative MS-based protein assays.