Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
2-Hydroxyglutarates Analysis by LCMS
Sigma-Aldrich has recently developed an application for the separation and identification of S-2-hydroxyglutaric acids and R-2-hydroxyglutaric acids by chiral LC-MS, using a CHIROBIOTIC® R Column (covalently bonded glycopeptide - ristocetin A). Chiral 2-hydroxyglutarates provide important molecular signatures of both healthy and diseased biological cells, their specific biochemical pathways, and inborn errors of metabolism.
The chiral differentiation and quantification of S-2-hydroxyglutaric acids and R-2-hydroxyglutaric acids is key for characterizing neurometabolic disorders like 2-hydroxyglutaric acidurias, which cause neurological impairment at a young age [1]. In human brain tumor patients mutations in the enzyme cytosolic isocitrate dehydrogenase 1 (IDH1) are found in approximately 80% of grade II-III gliomas and secondary glioblastomas. The demonstration that cancer-associated IDH1 mutations result in a new ability of the enzyme to catalyze the NADPH-dependent reduction of α-ketoglutarate to the oncometabolite R(−)-2-hydroxyglutarate [2] represents a milestone event in cancer biology. Cancer-associated IDH mutations in the IDH1 and IDH22 enzyme across gliomas, in addition to several hematologic malignanicies, have become of prognostic interest; as well as for biomarkers and therapeutic opportunities [3].
Figure 1. Hydroxyglutaric acid metabolism.
Figure 2. Cancer cell metabolism
Sigma-Aldrich provides the racemic and the pure chiral 2-hydroxyglutarates as well as the corresponding high-performance products and tools for their analysis.
For further information on isocitrate dehydrogenase (IDH) substrates, reaction products, assay kits, and isocitrate dehydrogenase in cancer metabolism, please view our IDH products for cancer metabolism.
To view our chiral separation offerings, visit sigma-aldrich.com/chiral
Figure 3. Method - Supelco® CHIROBIOTIC R, 5 μm particle size, L × I.D. 25 cm × 4.6 mm (13024AST) Mobile Phase: 75% EtOH/MeOH (3/1), 25% Water / 0.1% TEAA pH=4.5, Flow: 0.4 ml/min, Temp: 20° C. Upper chromatogram shows the separation of the racemate used to develop the method. Lower chromatogram depicts L-Hydroxy Glutaric Acid enantiomer purified from a real sample. Purity was determined to be 99.6% compared to D-Hydroxy Glutaric acid enantiomer, and identification was distinguished by optical rotation.
Figure 4. Racemate sample; MS scan of each enantiomer. Upper scan 9.00 - 10.60 min.; lower scan 10.61 - 12.38 min.
References:
[1] M.Kranendijk, E.A.Struys, G.S.Salomons, M.S.Van der Knaap, C. Jakobs, J.Inherit.Metab.Dis. 35, 571-587 (2012).
[2] L.Dang, D.W.White, S.Gross, B.D.Bennett, M.A.Bittinger, E.M.Driggers, V.R.Fantin, H.G.Jang, S.Jin, M.C.Keenan, K.M.Marks, R.M.Prins, P.S.Ward, K.E.Yen, L.M.Liau, J.D.Rabinowitz, L.C.Cantley, C.B.Thompson, M.G.Vander Heiden, S.M.Su, Nature 462, 739-744 (2009).
[3] K.E.Yen, M.A.Bittinger, S.M.Su, V.R.Fantin, Oncogene 29, 6409-6417 (2010).
Trademarks:
Supelco, and CHIROBIOTIC are registered trademarks of Sigma-Aldrich Co. LLC.
Related Links