Identification, Assay and Organic Impurity Profiling Methods for Aripiprazole following the United States Pharmacopeia Monograph
Aripiprazole USP Monograph Methods
Overview
Aripiprazole is an atypical antipsychotic and a partial dopamine agonist.
It is primarily used in the treatment of schizophrenia, bipolar disorder, major depressive disorder, tic disorders, and irritability associated with autism. Aripiprazole was first approved by the U.S. Food and DrugAdministration in November 2002 for schizophrenia and by the European Medicines Agency in June 2004 for acute manic and mixed episodes associated with bipolar disorder.
Common commercial brand Names:
Abilify and Aripiprex
Aripiprazole was developed by Otsuka in Japan. In the United States, Otsuka America markets it jointly with Bristol-Myers Squibb. In 2010, sales were $4.6 billion globally; the patent expired in 2015.
In this Compilation, we have used the USP 40–NF 35 experimental Conditions for Aripiprazole in the following Areas:
  • Identification — FTIR
  • Assay — HPLC (gradient method)
  • Related Substances — HPLC (gradient method)
The HPLC methods are gradient methods, thus they are nonscalable.
The same chromatographic conditions are used for methods in both the assay and related substances, and a full validation protocol can be found using these USP Reference Standards: USP Aripiprazole RS and USP Aripiprazole Related Compound F RS.
Identification and Assay
DEFINITION
Aripiprazole contains not less than (NLT) 98.0% and not more than (NMT) 102.0% of aripiprazole (C23H27Cl2N3O2), calculated on the dried basis.
IDENTIFICATION—FTIR <197K>
A. Infrared absorption
B. The retention time of the major peak of the Sample solution corresponds to that of the Standard solution, as obtained in the Assay.
ASSAY—HPLC (gradient method)
Procedure (protect the solutions from light):
Diluent: Acetonitrile, methanol, water, and acetic acid (30:10:60:1)
Solution A: Acetonitrile and 0.05% trifluoroacetic acid (10:90)
Solution B: Acetonitrile and 0.05% trifluoroacetic acid (90:10)
Gradient: See table.
    Detector: UV 254 nm
    Column: 4.6 mm × 10 cm (3 µm) packing L1
    Flow rate: 1.2 mL/min
    Injection volume: 20 µL
Note: The gradient was established on an HPLC system with a dwell volume of approximately 650 µL.
Time (min) Solution A (%) Solution B (%)
0 80 20
2 80 20
10 65 35
20 10 90
25 10 90
26 80 20
35 80 20
We have used a Purospher® STAR RP-18 endcapped (3 µm) 100 x 4.6 mm (Catalog No. 1.50469).
This is a gradient method and can therefore not be changed.
Assay Solutions and Analysis
System suitability solution: 1 µg/mL each of USP Aripiprazole and USP Aripiprazole Related Compound F in Diluent
Standard solution
: 0.1 mg/mL of USP Aripiprazole in Diluent
Sample solution
: 0.1 mg/mL of aripiprazole in Diluent
System Suitability
Samples: System suitability solution and Standard solution
Note: The relative retention times (RRT) for aripiprazole and aripiprazole related compound F are 1.0 and 1.1, respectively.
Suitability Requirements
  • Resolution: NLT 2.0 between aripiprazole and aripiprazole related compound F, System suitability solution
  • Tailing factor: NMT 1.5 for aripiprazole, System suitability solution
  • Relative standard deviation: NMT 1.0%, Standard solution
Analysis
Samples: Standard solution and Sample solution
Calculate the percentage of aripiprazole (C23H27Cl2N3O2) in the portion of the sample taken.
Result = (rU/rS) × (CS/CU) × 100
rU = peak area from the Sample solution
rS = peak area from the Standard solution
CS = concentration of USP Aripiprazole in the Standard solution (mg/mL)
CU = concentration of aripiprazole in the Sample solution (mg/mL)
Acceptance criteria: 98.0%–102.0% on the dried basis
Impurities Analysis
Inorganic Impurities
A. Residue on Ignition—USP General Chapter 281: NMT 0.1%
B. Heavy Metals, Method II—USP General Chapter 2311: NMT 10 ppm
Organic Impurities
(Protect the solutions from light.) Diluent, Solution A, Solution B, Mobile phase, System suitability solution, Standard solution, Sample solution, Chromatographic system, and System suitability: Proceed as directed in the Assay.
Analysis
Samples: Sample solution
Calculate the percentage of each impurity in the portion of aripiprazole taken.
Result = (ri/rU) × (1/F) × 100
ri = peak response of each impurity from the Sample solution
rU = peak response of aripiprazole from the Sample solution
F = relative response factor (see table)
USP Reference Standards
  • USP Aripiprazole
  • USP Aripiprazole Related Compound F
1Chapter valid until Jan. 1, 2018.
Related Substances (Organic Impurities)
Name Relative retention time (RRT)
Relative response factor (RRF) Acceptance criteria (NMT, %)
Aripiprazole related compound G2 0.9 0.72 0.10
Aripiprazole 1.0
Aripiprazole related compound F3,4 1.1 1.0
Aripiprazole 4,4-dimer5 1.3 1.0 0.10
Any other impurity 0.10
Total impurities 0.50
27-{4-[4-(2,3-Dichlorophenyl)piperazin-1-yl]butoxy}quinolin-2(1H)-one
34-(2,3-Dichlorophenyl)-1-[4-(2-oxo-1,2,3,4-tetrahydroquinolin-7-yloxy)butyl]piperazin 1-oxide
4For system suitability and identification purposes only
51,1¢-(Ethane-1,1-diyl)bis(2,3-dichloro-4-{4-[3,4-dihydroquinolin-2(1H)-one-7-yloxybutyl]piperazin-1-yl}benzene)
Identification Data
A. Infrared Absorption
The reference <197K> in a monograph signifies that the substance under examination is mixed intimately with potassium bromide. We recommend potassium bromide for IR spectroscopy—Uvasol® (Catalog No. 1.04907).
Assay and Related Substances Data
Chromatographic Conditions
Column: Purospher® STAR RP-18 endcapped (3 µm) 100 x 4.6 mm (Catalog No. 1.50469)
Injection: 20 µL
Detection: UV 254 nm
Cell: 10 µL
Flow rate: 1.2 mL/min
Mobile phase A: Acetonitrile and 0.05% trifluoroacetic acid (10:90 v/v)
Mobile phase B: Acetonitrile and 0.05% trifluoroacetic acid (90:10 v/v)
Gradient: See table.
Temperature: 40 °C
Diluent: Acetonitrile, methanol, water, and acetic acid (30:10:60:1 v/v)
Sample: 1 µg/mL (1 ppm) each of aripiprazole and aripiprazole related compound F in Diluent
Pressure drop: 95–170 Bar (1,377–2,465 psi)
Time (min) A (%) B (%)
0.01 80 20
2 80 20
10 65 35
20 10 90
25 10 90
26 80 20
35 80 20
Suitability Requirements
  • Resolution: NLT 2.0 between aripiprazole and aripiprazole related compound F
  • Tailing factor: NMT 1.5 for aripiprazole
  • Relative retention time (RRT):
        1.0 for aripiprazole and
        1.1 for aripiprazole related compound F
Chromatogram
Chromatographic Data (System Suitability Solution)
Compound Retention Time (min) RRT Tailing Factor Resolution
Aripiprazole 12.0 1.0 1.4
Aripiprazole related compound F 12.5 1.05 1.3 2.8
Chromatographic Data (100ppm Standard Solution)
Compound Retention Time (min) Tailing Factor Theoretical plates
Aripiprazole 12.0 1.4 16,118
Assay and Related Substances Validation Data
1. Specificity
Determined by injection of System suitability solution and determination of the retention time andrelative retention time for aripiprazole and aripiprazole related compound F.
Compound Retention time (min) RRT Tailing factor Resolution
Aripiprazole
12.0 1.0 1.4
Aripiprazole related compound F 12.5 1.05 1.3 2.8
2. Repeatability
Determined by injecting five samples with a Standard solution containing 100.0 ppm USP Aripiprazole and 1.0 ppm of USP Aripiprazole Related Compound F.
Standard solution Aripiprazole (area units)
Aripiprazole related compound F (area units)
1 60,114
6,047,450
2 60,363 6,080,108
3 60,308 6,086,256
4 60,174 6,081,386
5 60,316 6,089,267
Mean 60,255 6,076,893
STDEV 105.6 16,868.6
RSD (%) 0.18 0.28
3. Linearity, Limit of Detection (LOD), and Limit of Quantitation (LOQ)
Determined by injecting seven concentration levels from 0.1 to 1.5 ppm of USP Aripiprazole Related Compound F and nine concentration levels ranging from 1.0 to 150.0 ppm of aripiprazole.
Aripiprazole related compound F
LOD (ppb) LOQ (ppb)
7.6 23
Aripiprazole related compound F
(ppm) Area units
0.1 6,020
0.25 15,062
0.5 29,604
0.8 48,154
1.0 60,630
1.2 73,335
1.5 92,098
Aripiprazole
LOD (ppm) LOQ (ppm)
0.57
1.7
Aripiprazole
(ppm) Area units
1.0 63,167
5.0 292,127
10.0 586,674
25.0 1,587,549
50.0 3,013,178
80.0 4,878,836
100.0 6,076,893
120.0 7,401,807
150.0 9,241,987
3. Linearity, LOD, LOQ (cont.)
Determined by injecting seven concentration levels from 0.1 to 1.5 ppm of USP Aripiprazole Related Compound F and nine concentration levels ranging from 1.0 to 150.0 ppm of aripiprazole.
4. LOQ Accuracy
Determined by injecting 10 Standard solutions at LOQ level of USP Aripiprazole RS.
Injection Area units SD Area units RSD (%)
N=10 151,949 ±424.9 0.28
Recommended Products
  • Acetic acid (glacial) 100% anhydrous for analysis—EMSURE®  ACS, ISO, Reag. Ph. Eur. (Catalog No. 1.00063)
  • Acetonitrile (gradient grade for liquid chromatography)—LiChrosolv®  Reag. Ph. Eur. (Catalog No. 1.00030)
  • Methanol (gradient grade for liquid chromatography)—LiChrosolv®  Reag. Ph. Eur. (Catalog No. 1.06007)
  • Potassium bromide for IR spectroscopy—Uvasol®  (Catalog No. 1.04907)
  • Purospher®  STAR RP-18 endcapped (3 µm) 100 x 4.6 mm (Catalog No. 1.50469)
  • Trifluoroacetic acid for spectroscopy—Uvasol®  (Catalog No. 1.08262)
  • Water for chromatography (LC-MS grade)—LiChrosolv®  (Catalog No. 1.15333) or fresh water from the Milli-Q®  system
  • Aripiprazole United States Pharmacopeia (USP) Reference Standard (Catalog No. 1042634 USP)
  • Aripiprazole Related Compound F United States Pharmacopeia (USP) Reference Standard (Catalog No. 1042689 USP)
Materials