Ascentis® Express ES-Cyano Reversed Phase U/HPLC Columns
Stable, reversed phase packing for basic, acidic or neutral compounds
Ascentis® Express ES-Cyano HPLC column is a high-speed, high-performance liquid chromatography column based on 90 Å Fused-Core® particles. The Fused-Core® particle provides a thin porous shell of high-purity silica surrounding a solid silica core. This particle design exhibits very high column efficiency due to the shallow diffusion paths in the 0.5-micron thick porous shell and the small overall particle size of 2.7 µm. The sterically protected, extensively endcapped diisopropyl-cyanopropylsilane stationary phase of the Ascentis® Express ES-Cyano column provides a stable, reversed-phase packing that can be used for basic, acidic, or neutral compounds.
Ascentis® Express ES-Cyano Specifications
Silica: Type B (High purity silica)
Particle Platform: Superficially porous particles (SPP)
Phase Chemistry: Diisopropylcyanopropyl
Particle Size: 2.0 µm / 2.7 µm / 5.0 µm
Pore Size: 90 Å
Carbon Load: 3.4% / 3.5% / 2.5%
Surface Area: 120 m²/g / 135 m²/g /90 m²/g
pH Range: 2 - 8
Max Temperature at Low pH: 80 °C
Max Temperature at High pH: 40 °C
Endcapped: Yes
Reversed Phase ES-Cyano HPLC Columns with Fused-Core® Technology
Ascentis® Express ES-Cyano is a high-speed, high-performance liquid chromatography column based on a superficially porous Fused-Core® particle design.  The Fused-Core® particle provides a thin porous shell of high-purity silica surrounding a solid silica core.  This particle design exhibits very high column efficiency due to the shallow diffusion paths. 3 different particle sizes are available for your specific needs:
Fused-Core® particle size 2 µm
2.0 µm particles
Best Fused-Core UHPLC Column
An optimized solution for high-throughput small molecule analysis
Fused-Core® particle size 2.7 µm
2.7 µm particles
Fast HPLC on ANY System
A practical solution that delivers UHPLC performance from any HPLC
Fused-Core® particle size 5 µm
5.0 µm particles
The Lab Work-Horse Column
True plug and play solution for improving existing 3 or 5 µm fully porous particle HPLC columns
HPLC Guard Column
Materials