Ascentis ® Express HILIC U/HPLC Columns
HILIC for separation of polar compounds
HILIC chromatography is a variation of normal-phase chromatography with the distinction that one of the major components of the mobile phase is water. The high volatility of the mobile phase makes HILIC LC-MS friendly, and allows for increased MS sensitivity. Many classes of polar compounds can be retained on the Ascentis® Express HILIC column including polar neutrals, polar acids, and polar and non-polar basic amines. The HILIC column provides orthogonal selectivity to the Ascentis® Express C18, with the elution typically the opposite of that for C18. Non-polar compounds elute early and the highly polar compounds have greater retention and selectivity.
For high speed and high resolution separations on HPLC and UHPLC instruments, the  Fused-Core® particle technology of the Ascentis® Express HILIC column means that efficiency is typically 40% higher in comparison to fully porous particulate columns with same particle size.
Ascentis® Express HILIC Specifications
Silica: Type B (High purity silica)
Particle Platform: Superficially porous particles (SPP)
Phase Chemistry: bare silica
Particle Size: 2.0 µm / 2.7 µm / 5.0 µm
Pore Size: 90 Å
Carbon Load: unbonded
Surface Area: 120 m²/g / 135 m²/g / 90 m²/g
pH Range: 1 - 8
Max Temperature at Low pH: 60 °C
Max Temperature at High pH: 40 °C
Endcapped: na
HILIC Columns with Fused-Core ®Technology for Retention of Polar Compounds
Ascentis® Express HILIC is a high-speed, high-performance liquid chromatography column based on a superficially porous Fused-Core® particle design.  The Fused-Core® particle provides a thin porous shell of high-purity silica surrounding a solid silica core.  This particle design exhibits very high column efficiency due to the shallow diffusion paths. Three different particle sizes are available for your specific needs:
Fused-Core® particle size 2 µm
2.0 µm particles
Best Fused-Core UHPLC Column
An optimized solution for high-throughput small molecule analysis
Fused-Core® particle size 2.7 µm
2.7 µm particles
Fast HPLC on ANY System
A practical solution that delivers UHPLC performance from any HPLC
Fused-Core® particle size 5 µm
5.0 µm particles
The Lab Work-Horse Column
True plug and play solution for improving existing 3 or 5 µm fully porous particle HPLC columns
HPLC Guard Column
Materials