Ascentis® Express OH5 U/HPLC Columns
Pentahydroxy phase for fast analysis of polar compounds
Ascentis® Express OH5 is a high-speed, high-performance liquid chromatography column based on Fused-Core® particle technology. The OH5 phase is a highly polar ligand that possesses 5 hydroxyl groups tethered to the silica via novel proprietary linkage phase chemistry. This phase is designed to provide enhanced HILIC partitioning and limited ion exchange retention. This phase exhibits enhanced retention and performance based on the unique phase chemistry coupled with the Fused-Core® technology.
The Ascentis® Express OH5 HPLC column can be used for basic, acidic, zwitterionic or neutral polar compounds with unique selectivity in HILIC mode, which also allows for improved MS sensitivity due to the higher organic content of mobile phases in this mode of chromatography. It can be also be used for traditional normal phase separations using non-polar, totally organic mobile phases.
Ascentis® Express OH5 Specifications:
Silica: Type B (High purity silica)
Particle Platform: Superficially porous particles (SPP)
Phase Chemistry: Pentahydroxy
Particle Size: 2.0 µm / 2.7 µm /
5.0 µm
Pore Size: 90 Å
Carbon Load: 2.8% / 3.2% / 2.1%
Surface Area: 120 m²/g / 135 m²/g / 90 m²/g
pH Range: 2 - 9
Max Temperature at Low pH: 60 °C
Max Temperature at High pH: 40 °C
Endcapped: No
OH5 HPLC and UHPLC columns for traditional normal phase separations or HILIC
Ascentis® Express OH5 is a high-speed, high-performance liquid chromatography column based on a superficially porous Fused-Core® particle design.  The Fused-Core® particle provides a thin porous shell of high-purity silica surrounding a solid silica core.  This particle design exhibits very high column efficiency due to the shallow diffusion paths. Three different particle sizes are available for your specific needs:
Fused-Core® particle size 2 µm
2.0 µm particles
Best Fused-Core UHPLC Column
An optimized solution for high-throughput small molecule analysis
Fused-Core® particle size 2.7 µm
2.7 µm particles
Fast HPLC on ANY System
A practical solution that delivers UHPLC performance from any HPLC
Fused-Core® particle size 5 µm
5.0 µm particles
The Lab Work-Horse Column
True plug and play solution for improving existing 3 or 5 µm fully porous particle HPLC columns
HPLC Guard Column