D1691-17
Standard Test Methods for Zinc in Water
These test methods cover the determination of zinc in water. Section 26 on Quality Control pertains to these test methods. Two test methods are given as follows: Either dissolved or total recoverable zinc may be determined. These test methods have been used successfully with reagent grade water. See the specific test method for applicability to other matrices. It is the user’s responsibility to assure the validity of these test methods in other matrices. The values stated in SI units are to be regarded as standard. The values given in parentheses are mathematical conversion to inch-pound units that are provided for information only and are not considered standard.
Analysis Type (Application Method): Atomic absorption
Link to ASTM® D1691-17 Method
Compatible Filters:
Any 0.45 μm filter that will not contribute/remove metals. Examples are IC Millex® filter , or MCE or hydrophilic PTFE in vacuum filter holder. Forceps to handle filter membrane.