D2274:14
p-NITROANILINE
This test method provides a basis for the estimation of the oxidation stability of middle distillate fuels such as No. 2 fuel oil. The test method may not provide a prediction of the quantity of insolubles that will form in field storage over any given period of time. The amount of insolubles formed in such field storage is subject to the specific conditions which are too variable for this test method to predict accurately.
Analysis Type (Application Method): Gravimetric
Link to ASTM® D2274:14 Method
Compatible Filters:
Vacuum glassware plus accessories, 0.8 μm , 47 mm, matched weight MCE filters. Forceps to handle filter membrane.
Materials
Related Links
Categories