D3645:08
Standard Test Methods for Beryllium in Water
These test methods cover the determination of dissolved and total recoverable beryllium in most waters and wastewaters. The values stated in SI units are to be regarded as standard. No other units of measurement are included in this standard. Exposure to beryllium can affect the lungs and/or skin.
Analysis Type (Application Method): Atomic absorption
Link to ASTM® D3645:08 Method
Compatible Filters:
Vacuum glassware plus accessories. Forceps to handle filter membrane.