D3651:11
Standard Test Method for Barium in Brackish Water, Seawater, and Brines
This test method covers the determination of soluble barium ion in brackish water, sea-water, and brines by atomic absorption spectrophotometry. The actual working range of this test method is 1 to 5 mg/L barium. This test method was used successfully on artificial brine samples. It is the user′s responsibility to ensure the validity of this test method for waters of untested matrices.
Analysis Type (Application Method): Atomic absorption spectrophotometry (AAS)
Link to ASTM® D3651:11 Method
Compatible Filters:
Groundwater sampling capsules or portable vacuum filter holder with hand vacuum pump. Filtering in field at time of collection. Any 0.45 μm filter that will not contribute/remove metals. An example would the the IC Millex® filter. Forceps to handle filter membrane.