D3986:17
Standard Test Method for Measurement of Fluorides in Workplace Atmospheres by Ion-Selective Electrodes
This test method covers the determination of dissolved and total recoverable barium in brines, seawater, and brackish waters by direct-current argon plasma atomic emission spectroscopy (DCP–AES). This test method has been tested in the range from 10 mg/L to 20 mg/L. Samples shall be diluted to contain concentrations within the calibration range. Higher concentrations can also be determined by changing to a less sensitive emission line.
Analysis Type (Application Method): Atomic emission spectroscopy
Link to ASTM® D3986:17 Method
Compatible Filters:
Groundwater sampling capsules or portable vacuum filter holder with hand vacuum pump. Filtering in field at time of collection. Any 0.45 μm filter that will not contribute/remove metals. An example would be an IC Millex® filter. Forceps to handle filter membrane.