D3987-12
Standard Practice for Shake Extraction of Solid Waste with Water
This practice is intended as a rapid means for obtaining an extract of solid waste. The extract may be used to estimate the release of constituents of the solid waste under the laboratory conditions described in this procedure. This practice is not intended to provide an extract that is representative of the actual leachate produced from a solid waste in the field or to produce extracts to be used as the sole basis of engineering design.
Analysis Type (Application Method): Dry, shake, and vacuum or pressure filtration (extraction)
Link to ASTM® D3987-12 Method
Compatible Filters:
Various 47, 90 or 142 mm filtration products and filters, diameter and pressure or vacuum holder to be determined by the customer. Filters: 0.45 μm and 8 μm MCE filters, or material of choice, of appropriate diameter.. Forceps to handle filter membrane.
Materials
Related Links
Categories