D4185-17
Standard Test Method for Measurement of Metals in Workplace Atmospheres by Flame Atomic Absorption Spectrophotometry
This practice covers the collection, dissolution, and determination of trace metals in workplace atmospheres, by flame atomic absorption spectrophotometry. This standard does not purport to address all of the safety concerns, if any, associated with its use. It is the responsibility of the user of this standard to establish appropriate safety and health practices and determine the applicability of regulatory limitations prior to use.
Analysis Type (Application Method): AAS
Link to ASTM® D4185-17 Method
Compatible Filters:
25 or 37 mm 3 piece air Monitoring cassettes with 0.8 μm MCE membrane. Forceps to handle filter membrane.