D4765:13
Standard Test Method for Measurement of Fluorides in Workplace Atmospheres by Ion-Selective Electrodes
This test method covers the simultaneous collection and separate measurements of gaseous fluoride (for example, hydrogen fluoride) and particulate fluoride found in certain industrial workplaces. The gaseous fluorides and particulate fluorides collected are reported in terms of fluoride. The method covers sample collection, preparation, and fluoride measurement.
Analysis Type (Application Method): Fluoride ionselective electrode analysis
Link to ASTM® D4765:13 Method
Compatible Filters:
37 mm 0.8um MCE air Monitoring cassette and vacuum pump. Forceps to handle filter membrane.
Materials
Related Links
Categories