D4793-09 (2017)
Standard Test Method for Sequential Batch Extraction of Waste with Water
This test method is intended as a means for obtaining sequential extracts of a waste. This test method is a procedure for the sequential leaching of a waste containing at least five percent solids to generate solutions to be used to determine the constituents leached under the specified testing conditions. The extracts may be used to estimate the release of certain constituents of the waste under the laboratory conditions described in this test method.
Analysis Type (Application Method): Sequential batch extraction, pressure filtration
Link to ASTM® D4793-09 (2017) Method
Compatible Filters:
Pressure holder with sample chamber and filters. Forceps to handle filter membrane.
Materials
Related Links
Categories