D4807-05
p-NITROANILINE
This test method covers the determination of sediment in crude oils by membrane filtration. This test method has been validated for crude oils with sediments up to approximately 0.15 mass %. Measuring sediment content in crude oil is key in refinery operations and crude oil commerce.
Analysis Type (Application Method): Gravimetric
Link to ASTM® D4807-05 Method
Compatible Filters:
47 mm Vacuum glassware and accessories, 0.45 μm nylon filter. Forceps to handle filter membrane.
Materials
Related Links
Categories